3-Fluoro-2-iodo-5-methylphenol ≥97%

Supplier: AOBChem USA

Warning

34116-1G 34116-500MG 34116-5G 34116-25G
77469-764EA 284.28 USD
77469-764 77482-432 77492-854 77502-692
3-Fluoro-2-iodo-5-methylphenol ≥97%
3-Fluoro-2-iodo-5-methylphenol

Precaución: For research use only

Formula: C₇H₆FIO
MW: 252.025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403714
CAS Number: 2385718-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR