3-(9H-carbazol-9-yl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

34061-1G 34061-5G 34061-25G
77469-760EA 60.77 USD
77469-760 77492-850 77502-686
3-(9H-carbazol-9-yl)benzoic acid ≥97%
3-(9H-carbazol-9-yl)benzoic acid

Precaución: For research use only

Formula: C₁₉H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD34755949
CAS Number: 19287-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR