(2-Fluoro-5-methylphenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

340161-1G 340161-500MG 340161-5G 340161-25G
77469-752EA 415.09 USD
77469-752 77482-422 77492-842 77502-680
(2-Fluoro-5-methylphenyl)(propyl)sulfane ≥95%
(2-Fluoro-5-methylphenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₃FS
Storage Temperature: Ambient
CAS Number: 3002541-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR