(5-Bromo-2-fluoro-3-(methylthio)phenyl)(3-chlorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

340156-1G 340156-500MG 340156-5G 340156-25G
77469-748EA 415.09 USD
77469-748 77482-418 77492-838 77502-676
(5-Bromo-2-fluoro-3-(methylthio)phenyl)(3-chlorophenyl)methanol ≥95%
(5-Bromo-2-fluoro-3-(methylthio)phenyl)(3-chlorophenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₁BrClFOS
MW: 361.6569 g/mol
Storage Temperature: Ambient
CAS Number: 3002488-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR