(5-Bromo-2-fluoro-3-methoxyphenyl)(3-chlorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

340153-1G 340153-500MG 340153-5G 340153-25G
77469-744EA 415.09 USD
77469-744 77482-414 77492-834 77502-672
(5-Bromo-2-fluoro-3-methoxyphenyl)(3-chlorophenyl)methanol ≥95%
(5-Bromo-2-fluoro-3-methoxyphenyl)(3-chlorophenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₁BrClFO₂
MW: 345.5913 g/mol
Storage Temperature: Ambient
CAS Number: 3002512-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR