(5-Bromo-2-fluoro-3-methoxyphenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

340150-1G 340150-500MG 340150-5G 340150-25G
77469-740EA 415.09 USD
77469-740 77482-410 77492-830 77502-668
(5-Bromo-2-fluoro-3-methoxyphenyl)(propyl)sulfane ≥95%
(5-Bromo-2-fluoro-3-methoxyphenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₂BrFOS
MW: 279.1690 g/mol
Storage Temperature: Ambient
CAS Number: 3002477-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR