1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

340146-1G 340146-500MG 340146-5G 340146-25G
77469-736EA 415.09 USD
77469-736 77482-406 77492-826 77502-664
1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)propan-1-ol ≥95%
1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₀H₉F₅O
Storage Temperature: Ambient
CAS Number: 3002439-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR