(2,3-Difluoro-4-(trifluoromethyl)phenyl)(5-methylthiophen-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

340143-1G 340143-500MG 340143-5G 340143-25G
77469-732EA 415.09 USD
77469-732 77482-402 77492-822 77502-660
(2,3-Difluoro-4-(trifluoromethyl)phenyl)(5-methylthiophen-2-yl)methanol ≥95%
(2,3-Difluoro-4-(trifluoromethyl)phenyl)(5-methylthiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₃H₉F₅OS
MW: 308.2669 g/mol
Storage Temperature: Ambient
CAS Number: 3002499-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR