(2,3-Difluoro-4-(trifluoromethyl)phenyl)(isopropyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

340141-1G 340141-500MG 340141-5G 340141-25G
77469-728EA 415.09 USD
77469-728 77482-398 77492-818 77502-656
(2,3-Difluoro-4-(trifluoromethyl)phenyl)(isopropyl)sulfane ≥95%
(2,3-Difluoro-4-(trifluoromethyl)phenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₁₀H₉F₅S
Storage Temperature: Ambient
CAS Number: 3002512-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR