(3-Chlorophenyl)(2,3-difluoro-4-(trifluoromethyl)phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

340137-1G 340137-500MG 340137-5G 340137-25G
77469-724EA 415.09 USD
77469-724 77482-394 77492-814 77502-652
(3-Chlorophenyl)(2,3-difluoro-4-(trifluoromethyl)phenyl)methanol ≥95%
(3-Chlorophenyl)(2,3-difluoro-4-(trifluoromethyl)phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₈ClF₅O
Storage Temperature: Ambient
CAS Number: 3002541-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR