4-(2,3-Difluoro-4-(trifluoromethyl)phenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

340135-1G 340135-500MG 340135-5G 340135-25G
77469-720EA 415.09 USD
77469-720 77482-390 77492-810 77502-648
4-(2,3-Difluoro-4-(trifluoromethyl)phenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(2,3-Difluoro-4-(trifluoromethyl)phenyl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₂H₁₁F₅O₂
Storage Temperature: Ambient
CAS Number: 3002439-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR