5-Methoxybenzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

33996-1G 33996-500MG 33996-5G 33996-25G
77469-716EA 140.08 USD
77469-716 77482-386 77492-804 77502-640
5-Methoxybenzo[b]thiophene ≥95%
5-Methoxybenzo[b]thiophene

Precaución: For research use only

Formula: C₉H₈OS
MW: 164.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01860057
CAS Number: 20532-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR