4-Chloro-4'-fluorobenzophenone ≥95%

Supplier: AOBChem USA

Warning

339215-1G 339215-5G 339215-25G
77469-712EA 64.89 USD
77469-712 77492-800 77502-636
4-Chloro-4'-fluorobenzophenone ≥95%
4-Chloro-4'-fluorobenzophenone

Precaución: For research use only

Formula: C₁₃H₈ClFO
MW: 234.65 g/mol
Storage Temperature: Ambient
CAS Number: 2069-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR