2,4-Dichloro-3-fluorobenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

33884-1G 33884-500MG 33884-5G 33884-25G
77469-708EA 79.31 USD
77469-708 77482-380 77492-796 77502-632
2,4-Dichloro-3-fluorobenzyl alcohol ≥95%
2,4-Dichloro-3-fluorobenzyl alcohol

Precaución: For research use only

Formula: C₇H₅Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD28786031
CAS Number: 1615212-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR