3-(2-Bromo-3-fluoropyridin-4-yl)-2,4-dimethylpentan-3-ol ≥95%

Supplier: AOBChem USA

Warning

338743-1G 338743-500MG 338743-5G 338743-25G
77469-700EA 415.09 USD
77469-700 77482-372 77492-788 77502-624
3-(2-Bromo-3-fluoropyridin-4-yl)-2,4-dimethylpentan-3-ol ≥95%
3-(2-Bromo-3-fluoropyridin-4-yl)-2,4-dimethylpentan-3-ol

Precaución: For research use only

Formula: C₁₂H₁₇BrFNO
Storage Temperature: Ambient
CAS Number: 3002447-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR