2-(2-Bromo-3-fluoropyridin-4-yl)pentan-2-ol ≥95%

Supplier: AOBChem USA

Warning

338740-1G 338740-500MG 338740-5G 338740-25G
77469-696EA 415.09 USD
77469-696 77482-368 77492-784 77502-620
2-(2-Bromo-3-fluoropyridin-4-yl)pentan-2-ol ≥95%
2-(2-Bromo-3-fluoropyridin-4-yl)pentan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₃BrFNO
MW: 262.119 g/mol
Storage Temperature: Ambient
CAS Number: 3002487-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR