1-(2-Bromo-3-fluoropyridin-4-yl)-1-cyclopropylethanol ≥95%

Supplier: AOBChem USA

Warning

338735-1G 338735-500MG 338735-5G 338735-25G
77469-692EA 415.09 USD
77469-692 77482-364 77492-780 77502-616
1-(2-Bromo-3-fluoropyridin-4-yl)-1-cyclopropylethanol ≥95%
1-(2-Bromo-3-fluoropyridin-4-yl)-1-cyclopropylethanol

Precaución: For research use only

Formula: C₁₀H₁₁BrFNO
Storage Temperature: Ambient
CAS Number: 3002538-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR