1-(2-Bromo-3-fluoropyridin-4-yl)-2,2-dimethylpropan-1-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
338730-1G
338730-500MG
338730-5G
338730-25G
77469-688EA
415.09
USD
77469-688
77482-360
77492-776
77502-612
1-(2-Bromo-3-fluoropyridin-4-yl)-2,2-dimethylpropan-1-ol ≥95%
1-(2-Bromo-3-fluoropyridin-4-yl)-2,2-dimethylpropan-1-ol
Precaución: For research use only
Formula:
C₁₀H₁₃BrFNO Storage Temperature: Ambient |
CAS Number:
3002440-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...