1-(2-Bromo-3-fluoropyridin-4-yl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

338714-1G 338714-500MG 338714-5G 338714-25G
77469-684EA 415.09 USD
77469-684 77482-356 77492-772 77502-608
1-(2-Bromo-3-fluoropyridin-4-yl)-2-chloro-2,2-difluoroethanone ≥95%
1-(2-Bromo-3-fluoropyridin-4-yl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₇H₂BrClF₃NO
Storage Temperature: Ambient
CAS Number: 3002487-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR