1-(2-Fluoro-5-methyl-3-(methylthio)phenyl)butan-1-ol ≥95%

Supplier: AOBChem USA

Warning

338638-1G 338638-500MG 338638-5G 338638-25G
77469-680EA 415.09 USD
77469-680 77482-352 77492-768 77502-604
1-(2-Fluoro-5-methyl-3-(methylthio)phenyl)butan-1-ol ≥95%
1-(2-Fluoro-5-methyl-3-(methylthio)phenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₇FOS
MW: 228.326 g/mol
Storage Temperature: Ambient
CAS Number: 3002422-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR