(3,5-Di-tert-butylphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

338583-1G 338583-500MG 338583-5G 338583-25G
77469-672EA 415.09 USD
77469-672 77482-344 77492-760 77502-596
(3,5-Di-tert-butylphenyl)triethylsilane ≥95%
(3,5-Di-tert-butylphenyl)triethylsilane

Precaución: For research use only

Formula: C₂₀H₃₆Si
MW: 304.585 g/mol
Storage Temperature: Ambient
CAS Number: 2925590-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR