2-Chloro-1-(3,5-di-tert-butylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

338571-1G 338571-500MG 338571-5G 338571-25G
77469-668EA 415.09 USD
77469-668 77482-340 77492-756 77502-592
2-Chloro-1-(3,5-di-tert-butylphenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(3,5-di-tert-butylphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₆H₂₁ClF₂O
Storage Temperature: Ambient
CAS Number: 3002439-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR