(3,5-Di-tert-butylphenyl)(isopropyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

338569-1G 338569-500MG 338569-5G 338569-25G
77469-664EA 415.09 USD
77469-664 77482-336 77492-752 77502-588
(3,5-Di-tert-butylphenyl)(isopropyl)sulfane ≥95%
(3,5-Di-tert-butylphenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₁₇H₂₈S
Storage Temperature: Ambient
CAS Number: 3002456-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR