4-Chloro-3-fluoro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

33841-1G 33841-500MG 33841-5G 33841-25G
77469-660EA 426.42 USD
77469-660 77482-332 77492-748 77502-584
4-Chloro-3-fluoro-2-methylbenzoic acid ≥97%
4-Chloro-3-fluoro-2-methylbenzoic acid

Precaución: For research use only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20645712
CAS Number: 157652-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR