[3,5-Bis(1,1-dimethylethyl)phenyl]cyclohexylmethanone ≥95%

Supplier: AOBChem USA

Warning

338274-1G 338274-500MG 338274-5G 338274-25G
77469-656EA 415.09 USD
77469-656 77482-328 77492-744 77502-580
[3,5-Bis(1,1-dimethylethyl)phenyl]cyclohexylmethanone ≥95%
[3,5-Bis(1,1-dimethylethyl)phenyl]cyclohexylmethanone

Precaución: For research use only

Formula: C₂₁H₃₂O
MW: 300.48 g/mol
Storage Temperature: Ambient
CAS Number: 180740-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR