4-Chloro-2-fluoro-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33814-1G 33814-500MG 33814-5G 33814-25G
77469-652EA 131.84 USD
77469-652 77482-324 77492-740 77502-576
4-Chloro-2-fluoro-3-(methylthio)benzaldehyde ≥97%
4-Chloro-2-fluoro-3-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₈H₆ClFOS
MW: 204.649 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561330
CAS Number: 2145093-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR