4-Benzyloxy-2-formylphenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-(Benzyloxy)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

33717-1G 33717-5G 33717-25G
77469-648EA 60.77 USD
77469-648 77492-734 77502-570
4-Benzyloxy-2-formylphenylboronic acid pinacol ester ≥95%
4-Benzyloxy-2-formylphenylboronic acid pinacol ester

Precaución: For research use only

Formula: C₂₀H₂₃BO₄
MW: 338.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561321
CAS Number: 2695516-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR