6-Fluoro-2-hydroxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

33679-1G 33679-500MG 33679-5G
77469-644EA 576.8 USD
77469-644 77482-318 77492-730
6-Fluoro-2-hydroxy-3-methylbenzoic acid ≥97%
6-Fluoro-2-hydroxy-3-methylbenzoic acid

Precaución: For research use only

Formula: C₈H₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD22415140
CAS Number: 1783342-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR