(5-Bromo-2-fluorophenyl)(p-tolyl)methanol ≥95%

Supplier: AOBChem USA

Warning

335261-1G 335261-500MG 335261-5G 335261-25G
77469-632EA 415.09 USD
77469-632 77482-306 77492-718 77502-558
(5-Bromo-2-fluorophenyl)(p-tolyl)methanol ≥95%
(5-Bromo-2-fluorophenyl)(p-tolyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₂BrFO
MW: 295.1468 g/mol
Storage Temperature: Ambient
CAS Number: 1290728-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR