Methyl 2,6-difluoropyridine-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

33477-1G 33477-500MG 33477-5G
77469-628EA 628.3 USD
77469-628 77482-302 77492-714
Methyl 2,6-difluoropyridine-4-carboxylate ≥97%
Methyl 2,6-difluoropyridine-4-carboxylate

Precaución: For research use only

Formula: C₇H₅F₂NO₂
MW: 173.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698039
CAS Number: 119308-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR