3-Bromo-5-ethyl-2-fluorophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

33415-1G 33415-500MG 33415-5G
77469-624EA 580.92 USD
77469-624 77482-298 77492-710
3-Bromo-5-ethyl-2-fluorophenylboronic acid ≥95%
3-Bromo-5-ethyl-2-fluorophenylboronic acid

Precaución: For research use only

Formula: C₈H₉BBrFO₂
MW: 246.869 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30834055
CAS Number: 2121512-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR