(2-Fluoro-5-propoxyphenyl)(isopropyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

333847-1G 333847-500MG 333847-5G 333847-25G
77469-620EA 415.09 USD
77469-620 77482-294 77492-706 77502-550
(2-Fluoro-5-propoxyphenyl)(isopropyl)sulfane ≥95%
(2-Fluoro-5-propoxyphenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₁₂H₁₇FOS
Storage Temperature: Ambient
CAS Number: 3002506-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR