1,3-Dibromo-4-chloro-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

333827-1G 333827-500MG 333827-5G 333827-25G
77469-616EA 328.57 USD
77469-616 77482-290 77492-702 77502-546
1,3-Dibromo-4-chloro-2-fluorobenzene ≥95%
1,3-Dibromo-4-chloro-2-fluorobenzene

Precaución: For research use only

Formula: C₆H₂Br₂ClF
Storage Temperature: Ambient
CAS Number: 1521476-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR