(3-Bromo-6-chloro-2-fluorophenyl)(thiophen-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

333825-1G 333825-500MG 333825-5G 333825-25G
77469-612EA 415.09 USD
77469-612 77482-286 77492-698 77502-542
(3-Bromo-6-chloro-2-fluorophenyl)(thiophen-2-yl)methanol ≥95%
(3-Bromo-6-chloro-2-fluorophenyl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₁H₇BrClFOS
Storage Temperature: Ambient
CAS Number: 3002525-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR