(3-Bromo-6-chloro-2-fluorophenyl)(3-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

333823-1G 333823-500MG 333823-5G 333823-25G
77469-608EA 415.09 USD
77469-608 77482-282 77492-694 77502-538
(3-Bromo-6-chloro-2-fluorophenyl)(3-methoxyphenyl)methanol ≥95%
(3-Bromo-6-chloro-2-fluorophenyl)(3-methoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₁BrClFO₂
Storage Temperature: Ambient
CAS Number: 3002538-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR