Methyl-3,5-dichloroisonicotinate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 3,5-dichloropyridine-4-carboxylate

33381-1G 33381-500MG 33381-5G 33381-25G
77469-604EA 90.64 USD
77469-604 77482-278 77492-690 77502-534
Methyl-3,5-dichloroisonicotinate ≥95%
Methyl 3,5-dichloroisonicotinate

Precaución: For research use only

Formula: C₇H₅Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD06203945
CAS Number: 773874-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR