2,5-Difluoro-4-isopropoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33346-1G 33346-500MG 33346-5G 33346-25G
77469-600EA 109.18 USD
77469-600 77482-274 77492-686 77502-530
2,5-Difluoro-4-isopropoxyphenylboronic acid pinacol ester ≥97%
2,5-Difluoro-4-isopropoxyphenylboronic acid pinacol ester

Precaución: For research use only

Formula: C₁₅H₂₁BF₂O₃
MW: 298.133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30834050
CAS Number: 2121514-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR