(4-Ethoxyphenyl)(3-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

332836-1G 332836-500MG 332836-5G 332836-25G
77469-592EA 415.09 USD
77469-592 77482-266 77492-678 77502-524
(4-Ethoxyphenyl)(3-methoxyphenyl)methanol ≥95%
(4-Ethoxyphenyl)(3-methoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₈O₃
Storage Temperature: Ambient
CAS Number: 1226328-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR