2-Isopropoxyisonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

33238-1G 33238-500MG 33238-5G
77469-576EA 541.78 USD
77469-576 77482-250 77492-660
2-Isopropoxyisonicotinaldehyde ≥97%
2-Isopropoxyisonicotinaldehyde

Precaución: For research use only

Formula: C₉H₁₁NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28340492
CAS Number: 1462975-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR