(4-Bromo-1H-pyrazol-1-yl)acetonitrile ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-(4-Bromo-1H-pyrazol-1-yl)acetonitrile

33211-1G 33211-500MG 33211-5G 33211-25G
77469-572EA 79.31 USD
77469-572 77482-246 77492-656 77502-506
(4-Bromo-1H-pyrazol-1-yl)acetonitrile ≥95%
(4-Bromo-1H-pyrazol-1-yl)acetonitrile

Precaución: For research use only

Formula: C₅H₄BrN₃
MW: 186.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04969698
CAS Number: 925224-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR