2-Isopropyl-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33195-1G 33195-500MG 33195-5G
77469-568EA 484.1 USD
77469-568 77482-242 77492-652
2-Isopropyl-4-methoxybenzaldehyde ≥97%
2-Isopropyl-4-methoxybenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD24677639
CAS Number: 181035-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR