Pyridine, 2-phenyl-3-(trifluoromethyl)- ≥95%

Supplier: AOBChem USA

Warning

331820-1G 331820-500MG 331820-5G
77469-564EA 758.08 USD
77469-564 77482-238 77492-646
Pyridine, 2-phenyl-3-(trifluoromethyl)- ≥95%
Pyridine, 2-phenyl-3-(trifluoromethyl)-

Precaución: For research use only

Formula: C₁₂H₈F₃N
Storage Temperature: Ambient
CAS Number: 639782-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR