1-([2,2'-bithiophen]-5-yl)-2-chloro-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

331443-1G 331443-500MG 331443-5G 331443-25G
77469-560EA 415.09 USD
77469-560 77482-234 77492-642 77502-496
1-([2,2'-bithiophen]-5-yl)-2-chloro-2,2-difluoroethanone ≥95%
1-([2,2'-bithiophen]-5-yl)-2-chloro-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₅ClF₂OS₂
MW: 278.726 g/mol
Storage Temperature: Ambient
CAS Number: 3002498-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR