5-(Ethylthio)-2,2'-bithiophene ≥95%

Supplier: AOBChem USA

Warning

331440-1G 331440-500MG 331440-5G 331440-25G
77469-556EA 415.09 USD
77469-556 77482-230 77492-638 77502-492
5-(Ethylthio)-2,2'-bithiophene ≥95%
5-(Ethylthio)-2,2'-bithiophene

Precaución: For research use only

Formula: C₁₀H₁₀S₃
MW: 226.381 g/mol
Storage Temperature: Ambient
CAS Number: 215669-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR