(4-Isopropoxyphenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

331200-1G 331200-500MG 331200-5G 331200-25G
77469-552EA 415.09 USD
77469-552 77482-226 77492-634 77502-486
(4-Isopropoxyphenyl)(propyl)sulfane ≥95%
(4-Isopropoxyphenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₁₂H₁₈OS
Storage Temperature: Ambient
CAS Number: 3002455-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR