3-Chloro-4-methoxy-5-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

33119-1G 33119-500MG 33119-5G
77469-548EA 616.97 USD
77469-548 77482-222 77492-630
3-Chloro-4-methoxy-5-(trifluoromethyl)phenylboronic acid ≥97%
3-Chloro-4-methoxy-5-(trifluoromethyl)phenylboronic acid

Precaución: For research use only

Formula: C₈H₇BClF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD31382549
CAS Number: 2121515-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR