(4-Isobutoxyphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

331172-1G 331172-500MG 331172-5G 331172-25G
77469-544EA 415.09 USD
77469-544 77482-218 77492-624 77502-478
(4-Isobutoxyphenyl)trimethylsilane ≥95%
(4-Isobutoxyphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₃H₂₂OSi
MW: 222.399 g/mol
Storage Temperature: Ambient
CAS Number: 3002484-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR