4-Isobutoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

331168-1G 331168-500MG 331168-5G 331168-25G
77469-540EA 93.73 USD
77469-540 77482-214 77492-620 77502-474
4-Isobutoxybenzoic acid ≥95%
4-Isobutoxybenzoic acid

Precaución: For research use only

Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD01993659
CAS Number: 30762-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR