2-(4-isobutoxyphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

331158-1G 331158-500MG 331158-5G 331158-25G
77469-536EA 415.09 USD
77469-536 77482-210 77492-616 77502-470
2-(4-isobutoxyphenyl)propan-2-ol ≥95%
2-(4-isobutoxyphenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
MW: 208.297 g/mol
Storage Temperature: Ambient
CAS Number: 70309-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR