1-(4-Isobutoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

331152-1G 331152-5G 331152-25G
77469-532EA 60.77 USD
77469-532 77492-612 77502-466
1-(4-Isobutoxyphenyl)ethanone ≥95%
1-(4-Isobutoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00995652
CAS Number: 24242-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR